Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5H4orf47
Entrez Gene ID
102128187
Gene Name
chromosome 5 open reading frame, human C4orf47
Gene Aliases
-
Chromosome Location
Chr.5: 184840518 - 184864005 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005556434.2 XP_005556491.1 3 - 4 713 82
XM_005556436.2 XP_005556493.1 3 - 4 716 82

View Full Product Details