Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFP36
Entrez Gene ID
102141549
Gene Name
ZFP36 ring finger protein
Gene Aliases
-
Chromosome Location
Chr.19: 39910261 - 39912861 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005589176.2 XP_005589233.1 1 - 2 107 103

View Full Product Details