Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPS2
Entrez Gene ID
102125016
Gene Name
mitochondrial ribosomal protein S2
Gene Aliases
-
Chromosome Location
Chr.15: 2856258 - 2860000 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005580467.2 XP_005580524.1 1 - 2 120 70

View Full Product Details