Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPTE2
Entrez Gene ID
102136934
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2
Gene Aliases
-
Chromosome Location
Chr.17: 1874522 - 1971908 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015439049.1 XP_015294535.1 20 - 21 2013 109
GenBank mRNA AB169152.1 - 10 - 11 968 109

View Full Product Details