Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C3H7orf26
Entrez Gene ID
102144010
Gene Name
chromosome 3 open reading frame, human C7orf26
Gene Aliases
-
Chromosome Location
Chr.3: 41930088 - 41954013 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005549074.2 XP_005549131.1 5 - 6 1414 77

View Full Product Details