Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM71B
Entrez Gene ID
102142527
Gene Name
family with sequence similarity 71 member B
Gene Aliases
-
Chromosome Location
Chr.6: 156555429 - 156559458 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005558383.2 XP_005558440.1 1 - 2 707 67

View Full Product Details