Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPATA8
Entrez Gene ID
102125771
Gene Name
spermatogenesis associated 8
Gene Aliases
-
Chromosome Location
Chr.7: 77486987 - 77489152 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005560571.2 XP_005560628.1 2 - 3 463 96
XM_005560572.2 XP_005560629.1 2 - 3 459 96
XM_005560573.1 XP_005560630.1 2 - 3 212 96

View Full Product Details