Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBP5
Entrez Gene ID
102138977
Gene Name
retinol binding protein 5
Gene Aliases
-
Chromosome Location
Chr.11: 7556403 - 7562458 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005570005.2 XP_005570062.1 1 - 2 233 59

View Full Product Details