Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102125231
Entrez Gene ID
102125231
Gene Name
uncharacterized protein CXorf49-like
Gene Aliases
-
Chromosome Location
Chr.X: 68701622 - 68705518 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005593932.2 XP_005593989.1 5 - 6 1664 92

View Full Product Details