Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102132901
Entrez Gene ID
102132901
Gene Name
retinoic acid early transcript 1G protein
Gene Aliases
MIC, RAET1G
Chromosome Location
Chr.4: 55896419 - 55903900 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005595970.2 XP_005596027.2 1 - 2 174 97

View Full Product Details