Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102146276
Entrez Gene ID
102146276
Gene Name
zinc finger and SCAN domain-containing protein 5B
Gene Aliases
ZSCAN5B
Chromosome Location
Chr.19: 57009194 - 57012992 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015441075.1 XP_015296561.1 1 - 2 494 98

View Full Product Details