The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287634.1 | NP_001274563.1 | 3 - 3 | 599 | 114 |
GenBank mRNA | AB220385.1 | - | 3 - 3 | 599 | 114 |