Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bmp6
Entrez Gene ID
12161
Gene Name
bone morphogenetic protein 6
Gene Aliases
D13Wsu115e, Vgr1
Chromosome Location
Chr.13: 38345716 - 38499728 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007556.2 NP_031582.1 2 - 3 1061 122
GenBank mRNA AK041210.1 - - 1419 122
AK135770.1 - 2 - 3 694 122
BC138593.1 - 2 - 3 981 122
BC138595.1 - 2 - 3 981 122
J04566.1 - 2 - 3 643 122
X80992.1 - 2 - 3 1060 122

View Full Product Details