The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286226.1 | NP_001273155.1 | 1 - 2 | 450 | 57 |
NM_007581.3 | NP_031607.2 | 1 - 2 | 450 | 57 | |
GenBank mRNA | AF498245.1 | - | 1 - 2 | 48 | 57 |
AF498246.1 | - | 1 - 2 | 49 | 57 | |
BU709207.1 | - | 1 - 2 | 41 | 57 | |
CJ131971.1 | - | 1 - 2 | 189 | 57 | |
U20372.1 | - | 1 - 2 | 104 | 57 |