The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007635.4 | NP_031661.3 | 6 - 7 | 933 | 80 |
GenBank mRNA | AF005885.1 | - | 6 - 7 | 911 | 80 |
AK168891.1 | - | 5 - 6 | 776 | 80 | |
AK170651.1 | - | 6 - 7 | 917 | 80 | |
BC088980.1 | - | 6 - 7 | 739 | 80 | |
U95826.1 | - | 6 - 7 | 831 | 80 |