Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ephb6
Entrez Gene ID
13848
Gene Name
Eph receptor B6
Gene Aliases
Cekl, Mep
Chromosome Location
Chr.6: 41605040 - 41620507 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146351.1 NP_001139823.1 17 - 18 3450 78
NM_007680.4 NP_031706.3 17 - 18 3446 78
GenBank mRNA AK076442.1 - 17 - 18 3273 78
AK087423.1 - 17 - 18 3361 78
AK163823.1 - 8 - 9 1168 78
BC031924.1 - 16 - 17 3058 78
L77867.1 - 17 - 18 3140 78

View Full Product Details