The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290709.1 | NP_001277638.1 | 10 - 11 | 1119 | 60 |
NM_001290710.1 | NP_001277639.1 | 10 - 11 | 1092 | 60 | |
NM_001290711.1 | NP_001277640.1 | 10 - 11 | 1235 | 60 | |
NM_007897.3 | NP_031923.1 | 10 - 11 | 1116 | 60 | |
GenBank mRNA | AK031239.1 | - | 10 - 11 | 1114 | 60 |
AK035781.1 | - | 10 - 11 | 999 | 60 | |
AK137389.1 | - | 10 - 11 | 1157 | 60 | |
AK139021.1 | - | 10 - 11 | 1090 | 60 | |
AK141175.1 | - | 10 - 11 | 1119 | 60 | |
BC139362.1 | - | 10 - 11 | 1077 | 60 | |
BC139364.1 | - | 10 - 11 | 1077 | 60 | |
BC139765.1 | - | 3 - 4 | 568 | 60 | |
L12147.1 | - | 10 - 11 | 1082 | 60 |