The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008003.2 | NP_032029.1 | 2 - 3 | 502 | 65 |
GenBank mRNA | AF007268.1 | - | 2 - 3 | 502 | 65 |
AK017829.1 | - | 2 - 3 | 504 | 65 | |
BC021328.1 | - | 2 - 3 | 494 | 65 |