The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008190.1 | NP_032216.1 | 1 - 2 | 91 | 70 |
GenBank mRNA | AK075814.1 | - | 1 - 2 | 110 | 70 |
BC012640.1 | - | 1 - 2 | 102 | 70 | |
L05516.1 | - | 1 - 2 | 85 | 70 | |
M95175.1 | - | 1 - 2 | 91 | 70 |