The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008280.2 | NP_032306.2 | 8 - 9 | 1634 | 95 |
GenBank mRNA | AY228765.1 | - | 8 - 9 | 1386 | 95 |
BC021841.1 | - | 8 - 9 | 1634 | 95 | |
BC094050.1 | - | 8 - 9 | 1421 | 95 | |
X58426.1 | - | 8 - 9 | 1405 | 95 |