The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177973.1 | NP_001171444.1 | 7 - 8 | 1094 | 78 |
NM_001177974.1 | NP_001171445.1 | 7 - 8 | 1091 | 78 | |
NM_001177975.1 | NP_001171446.1 | 7 - 8 | 1091 | 78 | |
NM_001177976.1 | NP_001171447.1 | 7 - 8 | 1091 | 78 | |
NM_008363.2 | NP_032389.2 | 7 - 8 | 1091 | 78 | |
GenBank mRNA | AB088370.1 | - | 7 - 8 | 918 | 78 |
AF103876.1 | - | 7 - 8 | 1091 | 78 | |
AK045958.1 | - | 7 - 8 | 1000 | 78 | |
AK143694.1 | - | 7 - 8 | 1002 | 78 | |
AK144879.1 | - | 7 - 8 | 1000 | 78 | |
AK210148.1 | - | 3 - 4 | 358 | 78 | |
AY184362.1 | - | 7 - 8 | 1091 | 78 | |
AY184363.1 | - | 7 - 8 | 1091 | 78 | |
AY184364.1 | - | 7 - 8 | 1091 | 78 | |
BC004778.1 | - | 7 - 8 | 1086 | 78 | |
U56773.2 | - | 7 - 8 | 915 | 78 |