The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039537.2 | NP_001034626.1 | 2 - 3 | 509 | 77 |
NM_008501.2 | NP_032527.1 | 3 - 4 | 427 | 77 | |
GenBank mRNA | AF065917.1 | - | 2 - 3 | 219 | 77 |
AF065918.1 | - | 2 - 3 | 219 | 77 | |
AK042018.1 | - | 2 - 3 | 211 | 77 | |
AK155959.1 | - | 2 - 3 | 269 | 77 | |
BY209921.1 | - | 2 - 3 | 269 | 77 | |
X06381.1 | - | 1 - 2 | 77 | ||
X12810.1 | - | 2 - 3 | 77 |