Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Met
Entrez Gene ID
17295
Gene Name
met proto-oncogene
Gene Aliases
AI838057, HGF, HGFR, Par4, c-Met
Chromosome Location
Chr.6: 17463349 - 17573980 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008591.2 NP_032617.2 10 - 11 2617 106
GenBank mRNA AK148258.1 - 10 - 11 2470 106
BC117969.1 - 8 - 9 2261 106
BC117970.1 - 8 - 9 2257 106
CK625647.1 - 2 - 3 294 106
Y00671.1 - 8 - 9 2262 106

View Full Product Details