The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113198.1 | NP_001106669.1 | 7 - 8 | 1085 | 88 |
NM_001178049.1 | NP_001171520.1 | 7 - 8 | 972 | 88 | |
NM_008601.3 | NP_032627.1 | 6 - 7 | 776 | 88 | |
GenBank mRNA | AB221657.1 | - | 6 - 7 | 638 | 88 |
AK052895.1 | - | 7 - 8 | 944 | 88 | |
AK155350.1 | - | 7 - 8 | 1086 | 88 | |
BC108976.2 | - | 6 - 7 | 740 | 88 | |
BC108977.1 | - | 6 - 7 | 705 | 88 | |
Z23066.1 | - | 6 - 7 | 767 | 88 |