Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mybpc3
Entrez Gene ID
17868
Gene Name
myosin binding protein C, cardiac
Gene Aliases
-
Chromosome Location
Chr.2: 91118114 - 91136518 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008653.2 NP_032679.2 25 - 26 2639 98
GenBank mRNA AF097333.1 - 25 - 26 2590 98
AK146661.1 - 25 - 26 2640 98
AK146886.1 - 25 - 26 2640 98
AK169302.1 - 24 - 25 2617 98
AK169306.1 - 24 - 25 2617 98
BC094945.1 - 25 - 26 2636 98
BC131672.1 - 25 - 26 2615 98
BC154408.1 - 25 - 26 2660 98

View Full Product Details