The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008799.2 | NP_032825.2 | 2 - 3 | 549 | 75 |
GenBank mRNA | AK045985.1 | - | 2 - 3 | 550 | 75 |
AK182333.1 | - | 1 - 2 | 32 | 75 | |
DQ906042.1 | - | - | 529 | 75 | |
U10903.1 | - | 2 - 3 | 529 | 75 |