Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc17a1
Entrez Gene ID
20504
Gene Name
solute carrier family 17 (sodium phosphate), member 1
Gene Aliases
NAPI-1, Napi1, Npt1
Chromosome Location
Chr.13: 23867723 - 23895730 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170638.1 NP_001164109.1 7 - 8 854 78
NM_009198.3 NP_033224.3 7 - 8 946 78
NR_121611.1 7 - 8 1495 78
GenBank mRNA AK132800.1 - 7 - 8 1495 78
AK164712.1 - 7 - 8 856 78
BC013445.1 - 7 - 8 887 78
X77241.1 - 7 - 8 813 78

View Full Product Details