The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009275.4 | NP_033301.1 | 6 - 7 | 608 | 87 |
GenBank mRNA | AK004854.1 | - | 6 - 7 | 641 | 87 |
AK007859.1 | - | 6 - 7 | 614 | 87 | |
AK008383.1 | - | 6 - 7 | 651 | 87 | |
AK015469.1 | - | 6 - 7 | 673 | 87 | |
AK030437.1 | - | 6 - 7 | 609 | 87 | |
AK075583.1 | - | 6 - 7 | 611 | 87 | |
AK160600.1 | - | 5 - 6 | 1842 | 87 | |
AK167039.1 | - | 6 - 7 | 614 | 87 | |
BC003798.1 | - | 6 - 7 | 629 | 87 | |
U17343.1 | - | 6 - 7 | 622 | 87 |