The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009309.2 | NP_033335.1 | 5 - 6 | 838 | 86 |
GenBank mRNA | BC120807.1 | - | 5 - 6 | 810 | 86 |
X51683.1 | - | 5 - 6 | 838 | 86 |