The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009383.2 | NP_033409.1 | 2 - 3 | 488 | 86 |
GenBank mRNA | AK012085.1 | - | 2 - 3 | 132 | 86 |
U55861.1 | - | 2 - 3 | 264 | 86 |