Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ulk1
Entrez Gene ID
22241
Gene Name
unc-51 like kinase 1
Gene Aliases
AU041434, Unc51.1, mKIAA0722
Chromosome Location
Chr.5: 110784489 - 110810118 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009469.3 NP_033495.2 20 - 21 2428 70
GenBank mRNA AF053756.1 - 20 - 21 2120 70
AF072370.1 - 20 - 21 2111 70
AK122357.1 - 19 - 20 1930 70
AK148317.1 - 20 - 21 2421 70
AK154939.1 - 20 - 21 2445 70
AK156534.1 - 20 - 21 2405 70
BC053062.1 - 20 - 21 2088 70
BC057121.1 - 20 - 21 2428 70
BC059835.1 - 20 - 21 2397 70

View Full Product Details