The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110267.1 | NP_001103737.1 | 2 - 3 | 755 | 66 |
NM_001287057.1 | NP_001273986.1 | 5 - 6 | 1445 | 66 | |
NM_009505.4 | NP_033531.3 | 5 - 6 | 1445 | 66 | |
GenBank mRNA | BC061468.1 | - | 5 - 6 | 818 | 66 |
M95200.1 | - | 5 - 6 | 510 | 66 |