Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wnt11
Entrez Gene ID
22411
Gene Name
wingless-type MMTV integration site family, member 11
Gene Aliases
-
Chromosome Location
Chr.7: 98834921 - 98854747 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285792.1 NP_001272721.1 3 - 4 586 77
NM_001285795.1 NP_001272724.1 1 - 2 352 77
NM_009519.2 NP_033545.1 1 - 2 352 77
GenBank mRNA AK031573.1 - 1 - 2 195 77
AK077828.1 - 2 - 3 282 77
BC125482.1 - 1 - 2 299 77
BC125484.1 - 1 - 2 299 77
BC144947.1 - 1 - 2 299 77
X70800.1 - 1 - 2 359 77

View Full Product Details