The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009520.3 | NP_033546.2 | 1 - 2 | 407 | 118 |
GenBank mRNA | AF070988.1 | - | 1 - 2 | 284 | 118 |
AK035653.1 | - | 1 - 2 | 408 | 118 | |
AK043566.1 | - | 1 - 2 | 620 | 118 | |
BC119276.1 | - | 1 - 2 | 338 | 118 | |
BC119278.1 | - | 1 - 2 | 338 | 118 |