The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009553.2 | NP_033579.1 | 2 - 3 | 704 | 70 |
GenBank mRNA | AB221592.1 | - | 1 - 2 | 401 | 70 |
AK147779.1 | - | 2 - 3 | 600 | 70 | |
BC046961.1 | - | 2 - 3 | 704 | 70 | |
X55126.1 | - | 1 - 2 | 502 | 70 |