Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abcg1
Entrez Gene ID
11307
Gene Name
ATP-binding cassette, sub-family G (WHITE), member 1
Gene Aliases
AW413978, Abc8, White
Chromosome Location
Chr.17: 31057694 - 31117984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009593.2 NP_033723.1 14 - 15 1899 71
GenBank mRNA AF323659.1 - 14 - 15 1895 71
BC119471.2 - 14 - 15 1847 71
CO431055.1 - 2 - 3 144 71
U34920.1 - 14 - 15 1857 71
Z48745.1 - 14 - 15 1886 71

View Full Product Details