Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpd2
Entrez Gene ID
14571
Gene Name
glycerol phosphate dehydrogenase 2, mitochondrial
Gene Aliases
AA408484, AI448216, AU021455, AW494132, GPDH, Gdm1, Gpd-m, Gpdh-m, TISP38
Chromosome Location
Chr.2: 57237678 - 57370719 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145820.1 NP_001139292.1 9 - 10 1483 64
NM_010274.3 NP_034404.3 9 - 10 1358 64
GenBank mRNA AK034353.1 - 9 - 10 1267 64
AK144716.1 - 9 - 10 1484 64
AK151189.1 - 9 - 10 1271 64
AK167152.1 - 9 - 10 1296 64
BC021359.1 - 9 - 10 1452 64
D50430.1 - 9 - 10 1260 64
U60987.1 - 9 - 10 1294 64

View Full Product Details