The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111274.1 | NP_001104744.1 | 1 - 2 | 274 | 64 |
NM_001111276.1 | NP_001104746.1 | 1 - 2 | 274 | 64 | |
NM_184052.3 | NP_908941.1 | 1 - 2 | 274 | 64 | |
GenBank mRNA | AK038119.1 | - | 1 - 2 | 1357 | 64 |
X04480.1 | - | 1 - 2 | 136 | 64 | |
X04482.1 | - | 1 - 2 | 56 | 64 |