The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010548.2 | NP_034678.1 | 1 - 2 | 232 | 79 |
GenBank mRNA | AK152344.1 | - | 1 - 2 | 233 | 79 |
BC120612.1 | - | 1 - 2 | 207 | 79 | |
BC137844.1 | - | 1 - 2 | 207 | 79 | |
M37897.1 | - | 1 - 2 | 240 | 79 |