Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnk2
Entrez Gene ID
16526
Gene Name
potassium channel, subfamily K, member 2
Gene Aliases
A430027H14Rik, AI848635, TREK-1
Chromosome Location
Chr.1: 189207930 - 189402705 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010607.3 NP_034737.2 1 - 2 404 116
GenBank mRNA CJ163788.1 - 1 - 2 405 116
U73488.2 - 2 - 3 481 116

View Full Product Details