Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Matk
Entrez Gene ID
17179
Gene Name
megakaryocyte-associated tyrosine kinase
Gene Aliases
CHK, HYL, Ntk
Chromosome Location
Chr.10: 81252795 - 81262982 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010768.2 NP_034898.1 2 - 3 416 74
GenBank mRNA BC111864.1 - 1 - 2 94 74
L27738.1 - 2 - 3 170 74

View Full Product Details