The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011316.3 | NP_035446.1 | 1 - 2 | 112 | 80 |
GenBank mRNA | BC019212.1 | - | 1 - 2 | 148 | 80 |
U02554.1 | - | 1 - 2 | 143 | 80 |