The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011401.4 | NP_035531.3 | 3 - 4 | 626 | 87 |
GenBank mRNA | AK028041.1 | - | 3 - 4 | 400 | 87 |
AK045328.1 | - | 3 - 4 | 402 | 87 | |
AK155997.1 | - | 3 - 4 | 385 | 87 | |
AK164286.1 | - | 3 - 4 | 398 | 87 | |
BC034122.1 | - | 3 - 4 | 384 | 87 | |
BC058811.1 | - | 3 - 4 | 384 | 87 | |
CT010354.1 | - | 2 - 3 | 271 | 87 | |
M75135.1 | - | 3 - 4 | 350 | 87 | |
X61093.1 | - | 3 - 4 | 350 | 87 |