The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198977.1 | NP_001185906.1 | 1 - 2 | 173 | 80 |
GenBank mRNA | AK138741.1 | - | 1 - 2 | 108 | 80 |
BC065121.1 | - | 1 - 2 | 109 | 80 | |
Z49877.1 | - | 1 - 2 | 438 | 80 |