Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tap2
Entrez Gene ID
21355
Gene Name
transporter 2, ATP-binding cassette, sub-family B (MDR/TAP)
Gene Aliases
ABC18, AI462429, APT2, Abcb3, Ham-2, Ham2, MTP2, PSF2, RING11, Tap-2, Y1, jas
Chromosome Location
Chr.17: 34204085 - 34216321 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011530.3 NP_035660.3 11 - 12 2085 110
GenBank mRNA AK020022.1 - 10 - 11 2161 110
AK079667.1 - 11 - 12 1999 110
AK145234.1 - 11 - 12 2044 110
AK151376.1 - 11 - 12 1995 110
AK151483.1 - 11 - 12 1995 110
AK151657.1 - 11 - 12 2018 110
AK153287.1 - 11 - 12 1995 110
AK159200.1 - 11 - 12 2051 110
AK161091.1 - 11 - 12 2001 110
AK169645.1 - 8 - 9 1293 110
BC005578.1 - 11 - 12 1985 110
BC051257.1 - 11 - 12 1998 110
BC057668.1 - 11 - 12 2372 110
CT010387.1 - 10 - 11 1932 110
KJ650229.1 - 10 - 11 110
M90459.1 - 10 - 11 1932 110
U60087.1 - 10 - 11 1935 110
U60088.1 - 10 - 11 1935 110
U60089.1 - 10 - 11 1935 110
U60090.1 - 10 - 11 1935 110
U60091.1 - 10 - 11 1935 110
U60092.1 - 10 - 11 1932 110

View Full Product Details