The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253862.1 | NP_001240791.1 | 15 - 16 | 1603 | 71 |
NM_001253863.1 | NP_001240792.1 | 9 - 10 | 1054 | 71 | |
NM_001253864.1 | NP_001240793.1 | 8 - 9 | 982 | 71 | |
NM_001253865.1 | NP_001240794.1 | 10 - 11 | 1229 | 71 | |
NM_011544.3 | NP_035674.2 | 16 - 17 | 1732 | 71 | |
GenBank mRNA | AK132002.1 | - | 9 - 10 | 1042 | 71 |
AK134037.1 | - | 15 - 16 | 1604 | 71 | |
AK135376.1 | - | 8 - 9 | 950 | 71 | |
BC037097.1 | - | 10 - 11 | 1217 | 71 | |
M97635.1 | - | 16 - 17 | 1703 | 71 | |
M97636.1 | - | 1 - 2 | 1707 | 71 | |
X64840.1 | - | 16 - 17 | 1532 | 71 |