The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011571.3 | NP_035701.3 | 9 - 10 | 2152 | 88 |
GenBank mRNA | AB003494.1 | - | 9 - 10 | 2490 | 88 |
AK144302.1 | - | 9 - 10 | 1735 | 88 | |
BC099699.1 | - | 9 - 10 | 2152 | 88 |