The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011720.3 | NP_035850.2 | 3 - 4 | 369 | 59 |
GenBank mRNA | AF130349.1 | - | 3 - 4 | 371 | 59 |
AK050992.1 | - | 3 - 4 | 370 | 59 | |
BC119544.2 | - | 3 - 4 | 301 | 59 |