Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wif1
Entrez Gene ID
24117
Gene Name
Wnt inhibitory factor 1
Gene Aliases
AW107799, WIF-1
Chromosome Location
Chr.10: 121034004 - 121100642 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011915.2 NP_036045.1 9 - 10 1358 122
GenBank mRNA AF122923.1 - 9 - 10 1132 122
AK077698.1 - 9 - 10 1358 122
AK142443.1 - 9 - 10 1359 122
AK158359.1 - 9 - 10 1359 122
BC004048.1 - 5 - 6 638 122
BC013268.1 - 9 - 10 1159 122

View Full Product Details