Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ppp2r5c
- Entrez Gene ID
-
26931
- Gene Name
- protein phosphatase 2, regulatory subunit B', gamma
- Gene Aliases
-
2610043M05Rik, 2700063L20Rik, AI060890, AW545884, C85228, D12Bwg0916e, mKIAA0044
- Chromosome Location
-
Chr.12: 110447179 - 110583075 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK012612.1 |
- |
|
|
|
Chr. - - [-] on Build |
78 |
View Full Product Details